Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTGGTGGAGTCTCTTTCATTGC[A/T]GGGATCTTATGCTGGCAAAATCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC36 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC36 - leucine rich repeat containing 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161575.1 | 114 | Intron | NP_001155047.1 | |||
NM_018296.5 | 114 | Missense Mutation | CAG,CTG | Q,L 32 | NP_060766.5 | |
XM_005256025.2 | 114 | Missense Mutation | CAG,CTG | Q,L 32 | XP_005256082.1 | |
XM_005256026.2 | 114 | Missense Mutation | CAG,CTG | Q,L 32 | XP_005256083.1 | |
XM_005256027.2 | 114 | Missense Mutation | CAG,CTG | Q,L 32 | XP_005256084.1 | |
XM_005256028.1 | 114 | Intron | XP_005256085.1 | |||
XM_011523200.1 | 114 | Missense Mutation | CAG,CTG | Q,L 32 | XP_011521502.1 | |
XM_011523201.1 | 114 | Intron | XP_011521503.1 | |||
XM_011523202.2 | 114 | Intron | XP_011521504.1 | |||
XM_011523203.1 | 114 | Missense Mutation | CAG,CTG | Q,L 32 | XP_011521505.1 | |
XM_011523204.1 | 114 | Intron | XP_011521506.1 | |||
XM_011523205.1 | 114 | UTR 5 | XP_011521507.1 | |||
XM_011523206.2 | 114 | Missense Mutation | CAG,CTG | Q,L 32 | XP_011521508.1 | |
XM_017023400.1 | 114 | Missense Mutation | CAG,CTG | Q,L 32 | XP_016878889.1 | |
XM_017023401.1 | 114 | Missense Mutation | CAG,CTG | Q,L 32 | XP_016878890.1 | |
XM_017023402.1 | 114 | Missense Mutation | CAG,CTG | Q,L 32 | XP_016878891.1 |