Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCATGAACAGCAGCAGATTTGTC[C/G]TCAACGGAAAACAGCAGATCTTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605279 | ||||||||||||||||||||
Literature Links: |
CES3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CES3 - carboxylesterase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185176.1 | 410 | Intron | NP_001172105.1 | |||
NM_001185177.1 | 410 | Missense Mutation | CTC,GTC | L,V 111 | NP_001172106.1 | |
NM_024922.5 | 410 | Missense Mutation | CTC,GTC | L,V 111 | NP_079198.2 | |
XM_011522975.2 | 410 | Intron | XP_011521277.1 | |||
XM_011522976.2 | 410 | Missense Mutation | CTC,GTC | L,V 111 | XP_011521278.1 |