Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATGGGCCCCCTTACCAGCTCCCA[C/T]TCTTGGCTGTAGGGCTGCACGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610087 MIM: 605641 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRMT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRMT7 - protein arginine methyltransferase 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC7A6 - solute carrier family 7 member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC7A6OS - solute carrier family 7 member 6 opposite strand | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032178.2 | 708 | Silent Mutation | GAA,GAG | E,E 223 | NP_115554.2 | |
XM_011523372.2 | 708 | Silent Mutation | GAA,GAG | E,E 223 | XP_011521674.1 |