Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCACATAGTGCGCCTCAAGCCCA[C/G]CTGCTCTACAGACTCGTCGTTCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
IQCK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IQCK - IQ motif containing K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305121.1 | 749 | UTR 5 | NP_001292050.1 | |||
NM_001305122.1 | 749 | Missense Mutation | ACC,AGC | T,S 17 | NP_001292051.1 | |
NM_153208.2 | 749 | Missense Mutation | ACC,AGC | T,S 17 | NP_694940.1 | |
XM_011545736.2 | 749 | Intron | XP_011544038.1 | |||
XM_017022933.1 | 749 | Missense Mutation | ACC,AGC | T,S 17 | XP_016878422.1 |
KNOP1 - lysine rich nucleolar protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012991.2 | 749 | Intron | NP_001013009.2 | |||
XM_005255317.4 | 749 | Intron | XP_005255374.1 | |||
XM_006721046.3 | 749 | Intron | XP_006721109.1 | |||
XM_011545855.1 | 749 | Intron | XP_011544157.1 | |||
XM_017023231.1 | 749 | Intron | XP_016878720.1 | |||
XM_017023232.1 | 749 | Intron | XP_016878721.1 |