Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGGAAAAGCTCTAGAAATTCTT[C/A]TGGGAAGGAATAAGGCCTCTTGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616650 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSG1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GSG1L - GSG1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109763.1 | 1967 | UTR 3 | NP_001103233.1 | |||
NM_001323900.1 | 1967 | UTR 3 | NP_001310829.1 | |||
NM_001323901.1 | 1967 | UTR 3 | NP_001310830.1 | |||
NM_144675.2 | 1967 | UTR 3 | NP_653276.1 | |||
XM_011545750.2 | 1967 | Intron | XP_011544052.1 | |||
XM_011545751.2 | 1967 | Intron | XP_011544053.1 |
KIAA0556 - KIAA0556 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |