Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCAGCCGTGCGGCCTCCTCCAAC[A/G]TGCGCATGTCGCGCAGCACGGCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615700 MIM: 613288 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PYDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PYDC1 - pyrin domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152901.3 | 298 | Missense Mutation | ACG,ATG | T,M 78 | NP_690865.1 |
TRIM72 - tripartite motif containing 72 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008274.3 | 298 | Intron | NP_001008275.2 | |||
XM_017023254.1 | 298 | Intron | XP_016878743.1 |