Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCACTGGTGCCTGTGAGCCCATC[G/T]GCCTGCAGGGTGAGTCACCGCCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 151510 | ||||||||||||||||||||
Literature Links: |
ITGAX PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITGAX - integrin subunit alpha X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000887.4 | 359 | Missense Mutation | GGC,TGC | G,C 80 | NP_000878.2 | |
NM_001286375.1 | 359 | Missense Mutation | GGC,TGC | G,C 80 | NP_001273304.1 | |
XM_011545852.1 | 359 | Missense Mutation | GGC,TGC | G,C 80 | XP_011544154.1 | |
XM_011545854.1 | 359 | Missense Mutation | GGC,TGC | G,C 80 | XP_011544156.1 |