Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGCAGCTTCTCTCTTGGTGCGA[C/T]CTTTCCGGCCTCCACCTCCTGTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614812 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUPR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUPR1 - nuclear protein 1, transcriptional regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042483.1 | 398 | Missense Mutation | GAT,GGT | D,G 62 | NP_001035948.1 | |
NM_012385.2 | 398 | Missense Mutation | GAT,GGT | D,G 44 | NP_036517.1 |