Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCCATTGGTGCCAGCACTGGTT[C/T]CCCTTTACCTGAGCCAACAACCTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606248 MIM: 182160 | ||||||||||||||||||||
Literature Links: |
QPRT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
QPRT - quinolinate phosphoribosyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPN - sialophorin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030288.2 | 425 | Missense Mutation | TCC,TTC | S,F 87 | NP_001025459.1 | |
NM_003123.4 | 425 | Missense Mutation | TCC,TTC | S,F 87 | NP_003114.1 |