Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTATGAATAAGTCATCTACTTTC[C/T]TGAATCATGCTTCCACATAATATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603750 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AQP8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AQP8 - aquaporin 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZKSCAN2 - zinc finger with KRAB and SCAN domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012981.4 | 5361 | UTR 3 | NP_001012999.3 | |||
XM_017023200.1 | 5361 | UTR 3 | XP_016878689.1 |