Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATATTGTTTGCAGACAGTGACTGA[A/G]GATAACCCAGAGCTGCTGAAGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614951 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEATR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HEATR3 - HEAT repeat containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005256013.2 | 568 | Silent Mutation | GAA,GAG | E,E 130 | XP_005256070.1 | |
XM_011523188.2 | 568 | Silent Mutation | GAA,GAG | E,E 99 | XP_011521490.1 | |
XM_017023385.1 | 568 | UTR 5 | XP_016878874.1 | |||
XM_017023386.1 | 568 | UTR 5 | XP_016878875.1 |