Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACCGGGCCGTGGCTCTCACCCA[C/T]GCTGGACTCATCCGTCTGCAGGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614031 | ||||||||||||||||||||
Literature Links: |
GFOD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GFOD2 - glucose-fructose oxidoreductase domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RANBP10 - RAN binding protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320238.1 | 1101 | Missense Mutation | ATG,GTG | M,V 404 | NP_001307167.1 | |
NM_001320239.1 | 1101 | Missense Mutation | ATG,GTG | M,V 465 | NP_001307168.1 | |
NM_001320240.1 | 1101 | Missense Mutation | ATG,GTG | M,V 262 | NP_001307169.1 | |
NM_020850.2 | 1101 | Missense Mutation | ATG,GTG | M,V 491 | NP_065901.1 | |
XM_005256070.2 | 1101 | Missense Mutation | ATG,GTG | M,V 521 | XP_005256127.1 | |
XM_005256072.2 | 1101 | Missense Mutation | ATG,GTG | M,V 389 | XP_005256129.1 | |
XM_006721238.3 | 1101 | Missense Mutation | ATG,GTG | M,V 365 | XP_006721301.1 | |
XM_011523245.2 | 1101 | Missense Mutation | ATG,GTG | M,V 262 | XP_011521547.1 | |
XM_011523247.2 | 1101 | Missense Mutation | ATG,GTG | M,V 292 | XP_011521549.1 | |
XM_017023507.1 | 1101 | Missense Mutation | ATG,GTG | M,V 307 | XP_016878996.1 | |
XM_017023508.1 | 1101 | Missense Mutation | ATG,GTG | M,V 292 | XP_016878997.1 | |
XM_017023509.1 | 1101 | Missense Mutation | ATG,GTG | M,V 262 | XP_016878998.1 |