Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAATTCTTCCATCAACAAATATC[C/T]ACCTCTCCTGTCCAGCTTGCCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606005 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GGA2 - golgi associated, gamma adaptin ear containing, ARF binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015044.4 | 3105 | UTR 3 | NP_055859.1 | |||
XM_017023074.1 | 3105 | UTR 3 | XP_016878563.1 | |||
XM_017023075.1 | 3105 | UTR 3 | XP_016878564.1 |