Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCTGTTGAAGAGTTTGTTACCA[C/T]AGAGTCTGGCTTTCGAGTCATTCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611406 | ||||||||||||||||||||
Literature Links: |
DYNC1LI2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DYNC1LI2 - dynein cytoplasmic 1 light intermediate chain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286157.1 | 1475 | Missense Mutation | ATG,GTG | M,V 405 | NP_001273086.1 | |
NM_001323955.1 | 1475 | Missense Mutation | ATG,GTG | M,V 443 | NP_001310884.1 | |
NM_006141.2 | 1475 | Missense Mutation | ATG,GTG | M,V 482 | NP_006132.1 | |
XM_017023008.1 | 1475 | Intron | XP_016878497.1 |