Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGATCCCATTGTCACTCAGGGT[A/G]CACTGACTCCCAGGGTAGATTTCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611027 | ||||||||||||||||||||
Literature Links: |
SHCBP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SHCBP1 - SHC binding and spindle associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324318.1 | 1429 | Silent Mutation | TGC,TGT | C,C 462 | NP_001311247.1 | |
NM_001324319.1 | 1429 | Silent Mutation | TGC,TGT | C,C 422 | NP_001311248.1 | |
NM_024745.4 | 1429 | Silent Mutation | TGC,TGT | C,C 500 | NP_079021.3 | |
XM_011523335.2 | 1429 | Intron | XP_011521637.1 |