Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCATCCGTCAGGACCTTCCCGTA[C/T]AGAGCATCCAGCAGCCACTCAACGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137070 MIM: 606838 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FUS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FUS - FUS RNA binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PYCARD - PYD and CARD domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013258.4 | 437 | Silent Mutation | CTA,CTG | L,L 136 | NP_037390.2 | |
NM_145182.2 | 437 | Silent Mutation | CTA,CTG | L,L 117 | NP_660183.1 |
PYCARD-AS1 - PYCARD antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |