Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTCCTTCTCCAGCCAGTCACTGA[C/T]GTCCTGGGGCCCAGAGAGCTCCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604034 MIM: 602098 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERN2 - endoplasmic reticulum to nucleus signaling 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308220.1 | 2537 | Missense Mutation | ATC,GTC | I,V 742 | NP_001295149.2 | |
NM_033266.3 | 2537 | Missense Mutation | ATC,GTC | I,V 794 | NP_150296.4 | |
XM_011545708.1 | 2537 | Missense Mutation | ATC,GTC | I,V 650 | XP_011544010.1 | |
XM_011545709.1 | 2537 | Missense Mutation | ATC,GTC | I,V 650 | XP_011544011.1 | |
XM_011545711.2 | 2537 | Intron | XP_011544013.1 | |||
XM_011545712.2 | 2537 | Intron | XP_011544014.1 | |||
XM_011545713.1 | 2537 | Intron | XP_011544015.1 |
PLK1 - polo like kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |