Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAAGTTCACCATTGCTTCAAAAA[G/T]TTCCCTCATGCCCCCTTGCAGGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 121360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBFB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CBFB - core-binding factor, beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001755.2 | Intron | NP_001746.1 | ||||
NM_022845.2 | Intron | NP_074036.1 | ||||
XM_005256212.3 | Intron | XP_005256269.1 | ||||
XM_006721321.2 | Intron | XP_006721384.1 | ||||
XM_017023812.1 | Intron | XP_016879301.1 | ||||
XM_017023813.1 | Intron | XP_016879302.1 |