Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGGAATCCGCCATGTGTGGCTG[C/G]TGTCCTGCCAGGAGACAAGGCGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GSG1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSG1L - GSG1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109763.1 | 835 | Silent Mutation | CAC,CAG | H,Q 301 | NP_001103233.1 | |
NM_001323900.1 | 835 | Silent Mutation | CAC,CAG | H,Q 319 | NP_001310829.1 | |
NM_001323901.1 | 835 | Silent Mutation | CAC,CAG | H,Q 250 | NP_001310830.1 | |
NM_144675.2 | 835 | Silent Mutation | CAC,CAG | H,Q 146 | NP_653276.1 | |
XM_011545750.2 | 835 | Intron | XP_011544052.1 | |||
XM_011545751.2 | 835 | Intron | XP_011544053.1 |