Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATGATGGAGGGAAGCAGGCAGA[C/T]GCGAGTGTCTCGGCCATACAAGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
KLHL36 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLHL36 - kelch like family member 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303451.1 | 178 | Missense Mutation | ACG,ATG | T,M 8 | NP_001290380.1 | |
NM_024731.3 | 178 | Missense Mutation | ACG,ATG | T,M 8 | NP_079007.2 | |
XM_005256149.2 | 178 | Missense Mutation | ACG,ATG | T,M 8 | XP_005256206.1 | |
XM_017023674.1 | 178 | Missense Mutation | ACG,ATG | T,M 8 | XP_016879163.1 | |
XM_017023675.1 | 178 | Missense Mutation | ACG,ATG | T,M 8 | XP_016879164.1 |