Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCAACTGCAGACTGTAAATTTCC[A/G]ACAGAGAAAGGAATCTGTAGTTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611509 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CENPN - centromere protein N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100624.2 | 1064 | Missense Mutation | CAA,CGA | Q,R 42 | NP_001094094.2 | |
NM_001100625.2 | 1064 | Missense Mutation | CAA,CGA | Q,R 42 | NP_001094095.2 | |
NM_001270473.1 | 1064 | Missense Mutation | CAA,CGA | Q,R 42 | NP_001257402.1 | |
NM_001270474.1 | 1064 | Missense Mutation | CAA,CGA | Q,R 42 | NP_001257403.1 | |
NM_018455.5 | 1064 | Missense Mutation | CAA,CGA | Q,R 42 | NP_060925.2 | |
XM_006721236.3 | 1064 | Missense Mutation | CAA,CGA | Q,R 42 | XP_006721299.1 | |
XM_017023456.1 | 1064 | Missense Mutation | CAA,CGA | Q,R 42 | XP_016878945.1 |
CMC2 - C-X9-C motif containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |