Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGCCGCATTAGTTTCCCATTAAC[A/G]CTGAAGGCCTTTCCACACTCATTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194527 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF23 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF23 - zinc finger protein 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304492.1 | 2197 | Intron | NP_001291421.1 | |||
NM_001304493.1 | 2197 | Silent Mutation | AGC,AGT | S,S 400 | NP_001291422.1 | |
NM_001304494.1 | 2197 | Intron | NP_001291423.1 | |||
NM_145911.2 | 2197 | Silent Mutation | AGC,AGT | S,S 458 | NP_666016.1 |