Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCCGGTAGCCCAGCAGCTGCAG[C/T]GCACCAGCGCACAGTTCCTGCACGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605294 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHST6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHST6 - carbohydrate sulfotransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021615.4 | 1851 | Silent Mutation | GCA,GCG | A,A 352 | NP_067628.1 | |
XM_005255955.4 | 1851 | Silent Mutation | GCA,GCG | A,A 352 | XP_005256012.1 | |
XM_011523085.2 | 1851 | Silent Mutation | GCA,GCG | A,A 352 | XP_011521387.1 |