Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTTGAGGGCGATTCGATCGAAAG[C/T]TGGGTAACACCACCTTGTCCAGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603355 | ||||||||||||||||||||
Literature Links: |
MBTPS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MBTPS1 - membrane bound transcription factor peptidase, site 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003791.3 | 3424 | Missense Mutation | AAC,AGC | N,S 964 | NP_003782.1 | |
XM_017023816.1 | 3424 | Missense Mutation | AAC,AGC | N,S 523 | XP_016879305.1 | |
XM_017023817.1 | 3424 | Missense Mutation | AAC,AGC | N,S 468 | XP_016879306.1 |