Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609377 MIM: 610859 MIM: 607484 | ||||||||||||||||||||
Literature Links: |
ACD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACD - adrenocortical dysplasia homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C16orf86 - chromosome 16 open reading frame 86 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CARMIL2 - capping protein regulator and myosin 1 linker 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ENKD1 - enkurin domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PARD6A - par-6 family cell polarity regulator alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037281.1 | 828 | Nonsense Mutation | CGA,TGA | R,* 256 | NP_001032358.1 | |
NM_016948.2 | 828 | Nonsense Mutation | CGA,TGA | R,* 257 | NP_058644.1 | |
XM_005255977.3 | 828 | Nonsense Mutation | CGA,TGA | R,* 268 | XP_005256034.1 | |
XM_011523095.2 | 828 | Nonsense Mutation | CGA,TGA | R,* 267 | XP_011521397.1 | |
XM_011523096.2 | 828 | Nonsense Mutation | CGA,TGA | R,* 264 | XP_011521398.1 | |
XM_017023261.1 | 828 | Nonsense Mutation | CGA,TGA | R,* 252 | XP_016878750.1 |