Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGAGCGTTGGGGTGGCTTGGAAG[C/G]TGGTGTAGGTAGGTGGTGAGAGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612960 MIM: 602698 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESRP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESRP2 - epithelial splicing regulatory protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024939.2 | 1824 | Missense Mutation | ACC,AGC | T,S 579 | NP_079215.2 | |
XM_005256153.4 | 1824 | Missense Mutation | ACC,AGC | T,S 578 | XP_005256210.1 | |
XM_006721273.3 | 1824 | Missense Mutation | ACC,AGC | T,S 453 | XP_006721336.1 | |
XM_011523338.2 | 1824 | Missense Mutation | ACC,AGC | T,S 453 | XP_011521640.1 |
MIR6773 - microRNA 6773 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFATC3 - nuclear factor of activated T-cells 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |