Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCAGGGTGCAGGAGCTGCATCC[G/T]GAGAGCGGCGAAGAAGCGGTGACCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604191 | ||||||||||||||||||||
Literature Links: |
ZNF263 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF263 - zinc finger protein 263 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005741.4 | 377 | Silent Mutation | CCG,CCT | P,P 106 | NP_005732.2 | |
XM_006720831.1 | 377 | Silent Mutation | CCG,CCT | P,P 106 | XP_006720894.1 | |
XM_006720832.1 | 377 | Silent Mutation | CCG,CCT | P,P 106 | XP_006720895.1 | |
XM_006720833.1 | 377 | Silent Mutation | CCG,CCT | P,P 106 | XP_006720896.1 | |
XM_006720834.3 | 377 | UTR 5 | XP_006720897.1 | |||
XM_011522344.1 | 377 | Silent Mutation | CCG,CCT | P,P 106 | XP_011520646.1 | |
XM_017022850.1 | 377 | UTR 5 | XP_016878339.1 |