Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGCCGAGCCGCCGGGCGGCCAT[C/T]GAGCTGCTGATGGTTCTGGCGGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609077 MIM: 602438 MIM: 605235 MIM: 603500 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXL8 - F-box and leucine rich repeat protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018378.2 | 390 | Silent Mutation | ATC,ATT | I,I 90 | NP_060848.2 |
HSF4 - heat shock transcription factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040667.2 | 390 | Intron | NP_001035757.1 | |||
NM_001538.3 | 390 | Intron | NP_001529.2 |
NOL3 - nucleolar protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRADD - TNFRSF1A associated via death domain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |