Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCCATCGTCTTCATCGCTGTGG[A/G]CATCTATTTGTTGCTGCTGGGTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103850 MIM: 615175 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDOA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDOA - aldolase, fructose-bisphosphate A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000034.3 | 256 | UTR 5 | NP_000025.1 | |||
NM_001127617.2 | 256 | Intron | NP_001121089.1 | |||
NM_001243177.1 | 256 | Intron | NP_001230106.1 | |||
NM_184041.2 | 256 | Intron | NP_908930.1 | |||
NM_184043.2 | 256 | Intron | NP_908932.1 | |||
XM_011545768.2 | 256 | Intron | XP_011544070.1 |
FAM57B - family with sequence similarity 57 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |