Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCCACCGGGGGTCGGCTCCATGG[C/G]CAGCGCAGCCTGAAGCTCAGCTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608508 MIM: 603236 | ||||||||||||||||||||
Literature Links: |
CYBA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYBA - cytochrome b-245 alpha chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MVD - mevalonate diphosphate decarboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002461.2 | 1250 | Missense Mutation | CCC,GCC | P,A 359 | NP_002452.1 | |
XM_011523086.2 | 1250 | Missense Mutation | CCC,GCC | P,A 393 | XP_011521388.1 | |
XM_011523087.2 | 1250 | Missense Mutation | CCC,GCC | P,A 372 | XP_011521389.1 | |
XM_011523088.2 | 1250 | Missense Mutation | CCC,GCC | P,A 338 | XP_011521390.1 | |
XM_011523089.2 | 1250 | Missense Mutation | CCC,GCC | P,A 242 | XP_011521391.1 |