Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGCCATGCACGGCTTCTCGGTGA[A/C]CCTCCTGGGGTTCTGCAGTGAGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605907 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALG1 - ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019109.4 | 232 | Missense Mutation | AAC,ACC | N,T 64 | NP_061982.3 | |
XM_011522565.1 | 232 | Intron | XP_011520867.1 | |||
XM_017023457.1 | 232 | Missense Mutation | AAC,ACC | N,T 64 | XP_016878946.1 | |
XM_017023458.1 | 232 | UTR 5 | XP_016878947.1 |
C16orf89 - chromosome 16 open reading frame 89 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |