Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTCCCACACTGGCCCCCAGGGT[C/T]GCACGTTGGGTTAGTATTCTACTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 139311 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DKFZP434H168 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DKFZP434H168 - uncharacterized LOC26077 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GNAO1 - G protein subunit alpha o1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020988.2 | Intron | NP_066268.1 | ||||
NM_138736.2 | Intron | NP_620073.2 | ||||
XM_011523003.2 | Intron | XP_011521305.1 |
LOC283856 - uncharacterized LOC283856 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |