Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTAGCCCTGCAGTCAGGTCCTT[A/G]GGCCTGGCCTGGCCTCTCTGCCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608947 MIM: 616667 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASPHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASPHD1 - aspartate beta-hydroxylase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181718.3 | Intron | NP_859069.2 | ||||
XM_017023107.1 | Intron | XP_016878596.1 |
KCTD13 - potassium channel tetramerization domain containing 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178863.4 | Intron | NP_849194.1 | ||||
XM_011545783.2 | Intron | XP_011544085.1 | ||||
XM_011545784.2 | Intron | XP_011544086.1 | ||||
XM_017023104.1 | Intron | XP_016878593.1 | ||||
XM_017023105.1 | Intron | XP_016878594.1 | ||||
XM_017023106.1 | Intron | XP_016878595.1 |
SEZ6L2 - seizure related 6 homolog like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |