Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAATCTGCTTCTTTCTCCAGAT[A/C]ATAGATGATAAGGGCAACGTCCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614358 | ||||||||||||||||||||
Literature Links: |
ACSM2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACSM2A - acyl-CoA synthetase medium-chain family member 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308169.1 | Intron | NP_001295098.1 | ||||
NM_001308172.1 | Intron | NP_001295101.1 | ||||
NM_001308954.1 | Intron | NP_001295883.1 | ||||
XM_017022923.1 | Intron | XP_016878412.1 | ||||
XM_017022924.1 | Intron | XP_016878413.1 | ||||
XM_017022925.1 | Intron | XP_016878414.1 | ||||
XM_017022926.1 | Intron | XP_016878415.1 |