Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCAAAGGCGCCCTCCGGCAGAA[A/G]AACGTGCATGAGGTCAAGAACCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176970 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRKCB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRKCB - protein kinase C beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002738.6 | 284 | Silent Mutation | AAA,AAG | K,K 29 | NP_002729.2 | |
NM_212535.2 | 284 | Silent Mutation | AAA,AAG | K,K 29 | NP_997700.1 |