Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGCTGTGGCCTTGGCACAGAA[A/G]AATGTCAAGCTCTCAACAGAGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601051 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR662 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR662 - microRNA 662 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MSLN - mesothelin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177355.2 | 454 | Silent Mutation | AAA,AAG | K,K 92 | NP_001170826.1 | |
NM_005823.5 | 454 | Silent Mutation | AAA,AAG | K,K 92 | NP_005814.2 | |
NM_013404.4 | 454 | Silent Mutation | AAA,AAG | K,K 92 | NP_037536.2 | |
XM_005255034.4 | 454 | Silent Mutation | AAA,AAG | K,K 91 | XP_005255091.1 | |
XM_011522348.2 | 454 | Silent Mutation | AAA,AAG | K,K 92 | XP_011520650.1 | |
XM_017022853.1 | 454 | Silent Mutation | AAA,AAG | K,K 102 | XP_016878342.1 | |
XM_017022854.1 | 454 | Silent Mutation | AAA,AAG | K,K 101 | XP_016878343.1 | |
XM_017022855.1 | 454 | Silent Mutation | AAA,AAG | K,K 102 | XP_016878344.1 | |
XM_017022856.1 | 454 | Silent Mutation | AAA,AAG | K,K 101 | XP_016878345.1 | |
XM_017022857.1 | 454 | Silent Mutation | AAA,AAG | K,K 92 | XP_016878346.1 | |
XM_017022858.1 | 454 | Silent Mutation | AAA,AAG | K,K 91 | XP_016878347.1 |