Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGGACGTGGAAAAAAGTATTTG[A/G]GAGAAACAGGGAATGCTGGGAGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612081 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL34 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL34 - interleukin 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172771.1 | Intron | NP_001166242.1 | ||||
NM_001172772.1 | Intron | NP_001166243.1 | ||||
NM_152456.2 | Intron | NP_689669.2 | ||||
XM_011522902.2 | Intron | XP_011521204.1 | ||||
XM_011522903.2 | Intron | XP_011521205.1 |