Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGGGGCACTTCGCCTTTCAGAA[A/G]GTCATCATCTTCATCTTCCACTATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615367 MIM: 614244 MIM: 605121 | ||||||||||||||||||||
Literature Links: |
NTAN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NTAN1 - N-terminal asparagine amidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDXDC1 - pyridoxal dependent decarboxylase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001285444.1 | 1834 | Intron | NP_001272373.1 | |||
NM_001285445.1 | 1834 | Intron | NP_001272374.1 | |||
NM_001285447.1 | 1834 | Intron | NP_001272376.1 | |||
NM_001285448.1 | 1834 | Intron | NP_001272377.1 | |||
NM_001285449.1 | 1834 | Intron | NP_001272378.1 | |||
NM_001285450.1 | 1834 | Intron | NP_001272379.1 | |||
NM_001324019.1 | 1834 | Intron | NP_001310948.1 | |||
NM_001324020.1 | 1834 | Intron | NP_001310949.1 | |||
NM_001324021.1 | 1834 | Intron | NP_001310950.1 | |||
NM_015027.3 | 1834 | Intron | NP_055842.2 | |||
XM_005255173.1 | 1834 | Intron | XP_005255230.1 | |||
XM_005255176.2 | 1834 | Intron | XP_005255233.1 | |||
XM_006720865.2 | 1834 | Intron | XP_006720928.2 | |||
XM_017023059.1 | 1834 | Intron | XP_016878548.1 | |||
XM_017023060.1 | 1834 | Intron | XP_016878549.1 | |||
XM_017023061.1 | 1834 | Intron | XP_016878550.1 | |||
XM_017023062.1 | 1834 | Intron | XP_016878551.1 | |||
XM_017023063.1 | 1834 | Intron | XP_016878552.1 | |||
XM_017023064.1 | 1834 | Intron | XP_016878553.1 | |||
XM_017023065.1 | 1834 | Intron | XP_016878554.1 |
RRN3 - RRN3 homolog, RNA polymerase I transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301064.1 | 1834 | Missense Mutation | CTT,TTT | L,F 578 | NP_001287993.1 | |
NM_018427.4 | 1834 | Missense Mutation | CTT,TTT | L,F 608 | NP_060897.3 | |
XM_005255376.1 | 1834 | Missense Mutation | CTT,TTT | L,F 396 | XP_005255433.1 | |
XM_005255377.4 | 1834 | Missense Mutation | CTT,TTT | L,F 396 | XP_005255434.1 | |
XM_011522540.2 | 1834 | Missense Mutation | CTT,TTT | L,F 404 | XP_011520842.1 | |
XM_011522541.1 | 1834 | Missense Mutation | CTT,TTT | L,F 400 | XP_011520843.1 |