Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCCAGAGGACCAGAGGTTAATTT[A/T]TTCTGGGAAGCTGTTGTTGGATCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608070 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HERPUD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HERPUD1 - homocysteine inducible ER protein with ubiquitin like domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010989.2 | 328 | Missense Mutation | TAT,TTT | Y,F 58 | NP_001010989.1 | |
NM_001272103.1 | 328 | Missense Mutation | TAT,TTT | Y,F 58 | NP_001259032.1 | |
NM_014685.3 | 328 | Missense Mutation | TAT,TTT | Y,F 58 | NP_055500.1 |