Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGATCCTAAGGAAATATGTGTGG[A/G]TTCTTCTGACACATCCTACCACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613190 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAAF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAAF1 - dynein axonemal assembly factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318756.1 | 336 | Intron | NP_001305685.1 | |||
NM_178452.5 | 336 | Missense Mutation | GAT,GGT | D,G 52 | NP_848547.4 | |
XM_006721129.2 | 336 | Missense Mutation | GAT,GGT | D,G 52 | XP_006721192.1 | |
XM_011522853.2 | 336 | Missense Mutation | GAT,GGT | D,G 52 | XP_011521155.1 | |
XM_011522854.2 | 336 | Missense Mutation | GAT,GGT | D,G 52 | XP_011521156.1 | |
XM_011522855.2 | 336 | Missense Mutation | GAT,GGT | D,G 52 | XP_011521157.1 | |
XM_011522857.2 | 336 | Missense Mutation | GAT,GGT | D,G 52 | XP_011521159.1 | |
XM_011522858.2 | 336 | Missense Mutation | GAT,GGT | D,G 52 | XP_011521160.1 | |
XM_017022918.1 | 336 | Missense Mutation | GAT,GGT | D,G 52 | XP_016878407.1 | |
XM_017022919.1 | 336 | UTR 5 | XP_016878408.1 | |||
XM_017022920.1 | 336 | Intron | XP_016878409.1 | |||
XM_017022921.1 | 336 | Intron | XP_016878410.1 | |||
XM_017022922.1 | 336 | Intron | XP_016878411.1 |
HSDL1 - hydroxysteroid dehydrogenase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |