Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAGCCATGGGGCTCTCGTCCAG[C/G]CTGGAGGGCACTTCCACCTCTTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603246 MIM: 605383 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF3C1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GTF3C1 - general transcription factor IIIC subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286242.1 | 6069 | Silent Mutation | AGC,AGG | S,R 2049 | NP_001273171.1 | |
NM_001520.3 | 6069 | Silent Mutation | AGC,AGG | S,R 2074 | NP_001511.2 | |
XM_017023188.1 | 6069 | Silent Mutation | AGC,AGG | S,R 2011 | XP_016878677.1 |
IL21R - interleukin 21 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IL21R-AS1 - IL21R antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |