Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGCCTTCTCTCGCCAGATGCCG[A/G]TGCAGCTGACGACAGCCCTGCGTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613202 MIM: 602428 | ||||||||||||||||||||
Literature Links: |
CHTF8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHTF8 - chromosome transmission fidelity factor 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HAS3 - hyaluronan synthase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199280.1 | 98 | Missense Mutation | ATG,GTG | M,V 3 | NP_001186209.1 | |
NM_005329.2 | 98 | Missense Mutation | ATG,GTG | M,V 3 | NP_005320.2 | |
NM_138612.2 | 98 | Missense Mutation | ATG,GTG | M,V 3 | NP_619515.1 | |
XM_005255921.2 | 98 | Missense Mutation | ATG,GTG | M,V 3 | XP_005255978.1 | |
XM_011523061.2 | 98 | Missense Mutation | ATG,GTG | M,V 3 | XP_011521363.1 |