Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGCCCCTGCTCAGGCAGTCCTG[C/T]GCCCACTGCCAGCGAGTCCCTCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C16orf86 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C16orf86 - chromosome 16 open reading frame 86 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ENKD1 - enkurin domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GFOD2 - glucose-fructose oxidoreductase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243650.1 | 903 | Intron | NP_001230579.1 | |||
NM_030819.3 | 903 | Missense Mutation | ACA,GCA | T,A 289 | NP_110446.3 | |
XM_006721288.3 | 903 | Missense Mutation | ACA,GCA | T,A 184 | XP_006721351.1 | |
XM_017023738.1 | 903 | Missense Mutation | ACA,GCA | T,A 289 | XP_016879227.1 |