Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGACCACCATGGCTCCCAGGAA[A/G]GCAAAGACAGGAATGGTCTGGCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603355 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBTPS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MBTPS1 - membrane bound transcription factor peptidase, site 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003791.3 | 3551 | Silent Mutation | GCC,GCT | A,A 1006 | NP_003782.1 | |
XM_017023816.1 | 3551 | Silent Mutation | GCC,GCT | A,A 565 | XP_016879305.1 | |
XM_017023817.1 | 3551 | Silent Mutation | GCC,GCT | A,A 510 | XP_016879306.1 |