Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTAATGGAGTTCCTGCTCAATGT[C/G]ACCACAGCACCAGAATTTCGTCGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191329 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDZD9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDZD9 - PDZ domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173806.3 | 780 | Intron | NP_776167.2 | |||
XM_006721029.3 | 780 | Intron | XP_006721092.1 | |||
XM_011545785.2 | 780 | Intron | XP_011544087.1 | |||
XM_017023109.1 | 780 | Intron | XP_016878598.1 | |||
XM_017023110.1 | 780 | Intron | XP_016878599.1 |
UQCRC2 - ubiquinol-cytochrome c reductase core protein II | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003366.3 | 780 | Silent Mutation | GTC,GTG | V,V 140 | NP_003357.2 |