Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCAGGGATACGAGTAATGTTCC[A/G]TGCCCTTGTTCAGCAGCTGTGGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608333 MIM: 180663 | ||||||||||||||||||||
Literature Links: |
DOK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DOK4 - docking protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018110.3 | 1047 | Missense Mutation | ACG,ATG | T,M 252 | NP_060580.2 | |
XM_005256042.3 | 1047 | Missense Mutation | ACG,ATG | T,M 252 | XP_005256099.1 | |
XM_005256043.2 | 1047 | Missense Mutation | ACG,ATG | T,M 252 | XP_005256100.1 |
POLR2C - RNA polymerase II subunit C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |