Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTATGCCTCCCCGGCCATCCTG[C/T]TTCCCATGGAGCGGCAGCGCAACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600724 MIM: 610264 | ||||||||||||||||||||
Literature Links: |
CNGB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNGB1 - cyclic nucleotide gated channel beta 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEPP - testis, prostate and placenta expressed | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199046.2 | 275 | Missense Mutation | CTT,TTT | L,F 80 | NP_950247.2 | |
NM_199456.2 | 275 | Missense Mutation | CTT,TTT | L,F 80 | NP_955535.2 | |
XM_017023218.1 | 275 | Missense Mutation | CTT,TTT | L,F 25 | XP_016878707.1 |