Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTGCCGCATGGTCTCCACGCCC[A/T]CGGCGCCCACCTCCTCCTGCAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607490 MIM: 612060 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LDHD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LDHD - lactate dehydrogenase D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153486.3 | 1431 | Missense Mutation | GAG,GTG | E,V 483 | NP_705690.2 | |
NM_194436.2 | 1431 | Missense Mutation | GAG,GTG | E,V 460 | NP_919417.1 |
ZNRF1 - zinc and ring finger 1, E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |