Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGCCGGGAGGCACTTTGGATACC[A/G]CGTCCTGCAGGTCGGAAACGGGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 153370 | ||||||||||||||||||||
Literature Links: |
ITGAL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITGAL - integrin subunit alpha L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114380.1 | 319 | Missense Mutation | CAC,CGC | H,R 48 | NP_001107852.1 | |
NM_002209.2 | 319 | Missense Mutation | CAC,CGC | H,R 48 | NP_002200.2 | |
XM_005255313.1 | 319 | Missense Mutation | CAC,CGC | H,R 48 | XP_005255370.1 | |
XM_006721044.1 | 319 | Missense Mutation | CAC,CGC | H,R 48 | XP_006721107.1 | |
XM_011545849.1 | 319 | UTR 5 | XP_011544151.1 |