Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATTCCAATCAAAACACCAATGT[A/G]TAACTTTTGGAAACCTAAAGCAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603231 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR1F2P PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR1F2P - olfactory receptor family 1 subfamily F member 2 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF200 - zinc finger protein 200 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145446.1 | 2840 | UTR 3 | NP_001138918.1 | |||
NM_001145447.1 | 2840 | UTR 3 | NP_001138919.1 | |||
NM_001145448.1 | 2840 | UTR 3 | NP_001138920.1 | |||
NM_003454.3 | 2840 | UTR 3 | NP_003445.2 | |||
NM_198087.2 | 2840 | UTR 3 | NP_932353.1 | |||
NM_198088.2 | 2840 | UTR 3 | NP_932354.1 | |||
XM_005255556.1 | 2840 | UTR 3 | XP_005255613.1 | |||
XM_006720940.2 | 2840 | UTR 3 | XP_006721003.1 | |||
XM_006720941.2 | 2840 | UTR 3 | XP_006721004.1 | |||
XM_017023647.1 | 2840 | UTR 3 | XP_016879136.1 | |||
XM_017023648.1 | 2840 | Intron | XP_016879137.1 |